Assay Details
| Gene Symbol: NMU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628204.3 | XP_005628261.1 | 4 - 5 | 222 | 61 | |
| XM_005628205.3 | XP_005628262.1 | 4 - 5 | 222 | 61 | ||
| XM_014118849.2 | XP_013974324.1 | 5 - 6 | 406 | 61 | ||
| XM_022427001.1 | XP_022282709.1 | 4 - 5 | 222 | 61 | ||
| XM_022427002.1 | XP_022282710.1 | 5 - 6 | 323 | 61 | ||
More Information
Set Membership: |
|

NMU
Genomic Map