Assay Details
Gene Symbol: OSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022426424.1 | XP_022282132.1 | 2 - 3 | 539 | 132 | |
XM_022426425.1 | XP_022282133.1 | 2 - 3 | 573 | 132 | ||
XM_849897.4 | XP_854990.3 | 2 - 3 | 573 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
