Assay Details
| Gene Symbol: CARHSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003434858.3 | XP_003434906.1 | 3 - 4 | 432 | 69 | |
| XM_022420081.1 | XP_022275789.1 | 3 - 4 | 389 | 69 | ||
More Information
Set Membership: |
|

CARHSP1
Genomic Map