Assay Details
Gene Symbol: LSM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003432044.4 | XP_003432092.1 | 3 - 4 | 353 | 100 | |
XM_022403468.1 | XP_022259176.1 | 2 - 3 | 243 | 100 | ||
XM_850069.4 | XP_855162.2 | 2 - 3 | 268 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
