Assay Details
Gene Symbol: ADSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022422475.1 | XP_022278183.1 | 6 - 7 | 830 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
