Assay Details
| Gene Symbol: LOC607405 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619954.3 | XP_005620011.1 | 2 - 3 | 1196 | 74 | |
| XM_022418478.1 | XP_022274186.1 | 1 - 2 | 409 | 74 | ||
| XM_851891.5 | XP_856984.1 | 3 - 4 | 1269 | 74 | ||
More Information
Set Membership: |
|

LOC607405
Genomic Map