Assay Details
Gene Symbol: STYXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620982.3 | XP_005621039.1 | 531 | 72 | ||
XM_005620983.2 | XP_005621040.1 | 649 | 72 | |||
XM_005620984.3 | XP_005621041.1 | 334 | 72 | |||
XM_005620985.2 | XP_005621042.1 | 606 | 72 | |||
XM_014114178.2 | XP_013969653.1 | 470 | 72 | |||
XM_022419584.1 | XP_022275292.1 | 412 | 72 | |||
XM_022419585.1 | XP_022275293.1 | 407 | 72 | |||
XM_022419586.1 | XP_022275294.1 | 392 | 72 | |||
XM_844156.4 | XP_849249.1 | 624 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
