Assay Details
Gene Symbol: LOC100856570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003640075.4 | XP_003640123.1 | 9 - 10 | 1459 | 98 | |
XM_005638844.3 | XP_005638901.1 | 9 - 10 | 1045 | 98 |
Gene Symbol: LOC487740 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014109719.2 | XP_013965194.1 | 9 - 10 | 1045 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
