Assay Details
| Gene Symbol: UBE2D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022408334.1 | XP_022264042.1 | 422 | 91 | ||
| XM_850890.5 | XP_855983.1 | 600 | 91 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: UBE2D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022408334.1 | XP_022264042.1 | 422 | 91 | ||
| XM_850890.5 | XP_855983.1 | 600 | 91 | |||
Set Membership: |
|