Assay Details
Gene Symbol: PAIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014107449.2 | XP_013962924.1 | 472 | 115 | ||
XM_014107452.2 | XP_013962927.1 | 545 | 115 | |||
XM_014107453.2 | XP_013962928.1 | 461 | 115 | |||
XM_014107455.2 | XP_013962930.1 | 472 | 115 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
