Assay Details
| Gene Symbol: NARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014109219.2 | XP_013964694.1 | 293 | 116 | ||
| XM_843967.5 | XP_849060.3 | 306 | 116 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: NARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014109219.2 | XP_013964694.1 | 293 | 116 | ||
| XM_843967.5 | XP_849060.3 | 306 | 116 | |||
Set Membership: |
|