Assay Details
| Gene Symbol: RAB11FIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003432094.3 | XP_003432142.1 | 1789 | 89 | ||
| XM_005629868.3 | XP_005629925.1 | 3457 | 89 | |||
| XM_005629869.3 | XP_005629926.1 | 3381 | 89 | |||
| XM_022404057.1 | XP_022259765.1 | 3722 | 89 | |||
| XM_539967.5 | XP_539967.2 | 3664 | 89 | |||
More Information
Set Membership: |
|

RAB11FIP1
Genomic Map