Assay Details
| Gene Symbol: IGBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641443.3 | XP_005641500.2 | 4 - 5 | 879 | 94 | |
| XM_022415678.1 | XP_022271386.1 | 3 - 4 | 587 | 94 | ||
More Information
Set Membership: |
|

IGBP1
Genomic Map