Assay Details
Gene Symbol: SYNE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014115793.2 | XP_013971268.1 | 3 - 4 | 616 | 56 | |
XM_014115808.2 | XP_013971283.1 | 3 - 4 | 616 | 56 | ||
XM_022404162.1 | XP_022259870.1 | 3 - 4 | 616 | 56 | ||
XM_022404168.1 | XP_022259876.1 | 3 - 4 | 616 | 56 | ||
XM_022404172.1 | XP_022259880.1 | 3 - 4 | 616 | 56 | ||
XM_022404182.1 | XP_022259890.1 | 3 - 4 | 616 | 56 | ||
XM_022404191.1 | XP_022259899.1 | 3 - 4 | 616 | 56 | ||
XM_022404202.1 | XP_022259910.1 | 3 - 4 | 616 | 56 | ||
XM_022404211.1 | XP_022259919.1 | 3 - 4 | 616 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
