Assay Details
| Gene Symbol: RPL35A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022413845.1 | XP_022269553.1 | 4 - 5 | 363 | 89 | |
| XM_535773.5 | XP_535773.1 | 4 - 5 | 428 | 89 | ||
More Information
Set Membership: |
|

RPL35A
Genomic Map