Assay Details
| Gene Symbol: VAPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014115582.2 | XP_013971057.1 | 2 - 3 | 313 | 139 | |
| XM_014115583.2 | XP_013971058.1 | 3 - 4 | 283 | 139 | ||
| XM_022421636.1 | XP_022277344.1 | 3 - 4 | 283 | 139 | ||
| XM_854674.5 | XP_859767.1 | 2 - 3 | 286 | 139 | ||
More Information
Set Membership: |
|

VAPA
Genomic Map