Assay Details
| Gene Symbol: BCL7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_543381.6 | XP_543381.2 | 541 | 116 | ||
| XM_854760.5 | XP_859853.1 | 540 | 116 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: BCL7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_543381.6 | XP_543381.2 | 541 | 116 | ||
| XM_854760.5 | XP_859853.1 | 540 | 116 | |||
Set Membership: |
|