Assay Details
| Gene Symbol: SLX1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621358.3 | XP_005621415.1 | 1 - 2 | 220 | 94 | |
| XM_844501.5 | XP_849594.1 | 1 - 2 | 220 | 94 | ||
More Information
Set Membership: |
|

SLX1A
Genomic Map