Assay Details
| Gene Symbol: MAP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022410341.1 | XP_022266049.1 | 4 - 5 | 6861 | 63 | |
| XM_535269.5 | XP_535269.4 | 5 - 6 | 7175 | 63 | ||
More Information
Set Membership: |
|

MAP1B
Genomic Map