Assay Details
| Gene Symbol: GPR89A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630713.3 | XP_005630770.1 | 4 - 5 | 481 | 103 | |
| XM_022404930.1 | XP_022260638.1 | 4 - 5 | 483 | 103 | ||
| XM_022404931.1 | XP_022260639.1 | 4 - 5 | 481 | 103 | ||
| XM_022404932.1 | XP_022260640.1 | 3 - 4 | 377 | 103 | ||
| XM_844483.5 | XP_849576.1 | 4 - 5 | 481 | 103 | ||
More Information
Set Membership: |
|

GPR89A
Genomic Map