Assay Details
Gene Symbol: ZNF608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005626461.3 | XP_005626518.1 | 8 - 9 | 5302 | 70 | |
XM_005626464.3 | XP_005626521.1 | 7 - 8 | 3625 | 70 | ||
XM_005626465.3 | XP_005626522.1 | 7 - 8 | 4008 | 70 | ||
XM_014117686.2 | XP_013973161.1 | 9 - 10 | 5328 | 70 | ||
XM_014117687.2 | XP_013973162.1 | 8 - 9 | 4908 | 70 | ||
XM_022425226.1 | XP_022280934.1 | 9 - 10 | 4996 | 70 | ||
XM_022425227.1 | XP_022280935.1 | 8 - 9 | 5302 | 70 | ||
XM_538605.6 | XP_538605.2 | 8 - 9 | 5113 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
