Assay Details
| Gene Symbol: LOC610164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_847582.3 | XP_852675.1 | 2 - 3 | 467 | 146 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC610164| Gene Symbol: LOC610164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_847582.3 | XP_852675.1 | 2 - 3 | 467 | 146 | |
Set Membership: |
|