Assay Details
| Gene Symbol: GSPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621531.1 | XP_005621588.1 | 10 - 11 | 1222 | 122 | |
| XM_005621532.3 | XP_005621589.1 | 9 - 10 | 1017 | 122 | ||
| XM_022420056.1 | XP_022275764.1 | 10 - 11 | 1024 | 122 | ||
| XM_536971.6 | XP_536971.1 | 10 - 11 | 1066 | 122 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GSPT1
Genomic Map