Assay Details
| Gene Symbol: NDUFS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252295.1 | NP_001239224.1 | 5 - 6 | 407 | 128 | |
| XM_005631068.3 | XP_005631125.1 | 5 - 6 | 455 | 128 | ||
| XM_005631069.3 | XP_005631126.1 | 5 - 6 | 393 | 128 | ||
| XM_005631070.3 | XP_005631127.1 | 5 - 6 | 496 | 128 | ||
| XM_014120878.2 | XP_013976353.1 | 5 - 6 | 453 | 128 | ||
| GenBank mRNA | CO682700.1 | - | 1 | 307 | 128 | |
| DN411417.1 | - | 5 - 6 | 407 | 128 | ||
More Information
Set Membership: |
|

NDUFS8
Genomic Map