Assay Details
Gene Symbol: WASHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014117392.2 | XP_013972867.1 | 15 - 16 | 1532 | 105 | |
XM_531762.6 | XP_531762.3 | 15 - 16 | 1532 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
