Assay Details
| Gene Symbol: THNSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630460.3 | XP_005630517.1 | 7 - 8 | 1664 | 148 | |
| XM_005630461.3 | XP_005630518.1 | 7 - 8 | 1887 | 148 | ||
| XM_014120508.2 | XP_013975983.1 | 6 - 7 | 2603 | 148 | ||
| XM_532965.5 | XP_532965.2 | 7 - 8 | 1419 | 148 | ||
More Information
Set Membership: |
|

THNSL2
Genomic Map