Assay Details
| Gene Symbol: NOX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103218.1 | NP_001096688.1 | 5 - 6 | 828 | 106 | |
| XM_022412487.1 | XP_022268195.1 | 5 - 6 | 1263 | 106 | ||
| XM_022412488.1 | XP_022268196.1 | 5 - 6 | 846 | 106 | ||
| XM_022412489.1 | XP_022268197.1 | 5 - 6 | 1227 | 106 | ||
| XM_022412490.1 | XP_022268198.1 | 5 - 6 | 850 | 106 | ||
More Information
Set Membership: |
|

NOX5
Genomic Map