Assay Details
Gene Symbol: CADM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619738.3 | XP_005619795.1 | 610 | 140 | ||
XM_014113403.2 | XP_013968878.1 | 609 | 140 | |||
XM_014113406.2 | XP_013968881.1 | 609 | 140 | |||
XM_014113407.2 | XP_013968882.1 | 609 | 140 | |||
XM_022418268.1 | XP_022273976.1 | 1013 | 140 | |||
XM_022418269.1 | XP_022273977.1 | 1011 | 140 | |||
XM_022418271.1 | XP_022273979.1 | 1013 | 140 | |||
XM_022418272.1 | XP_022273980.1 | 1011 | 140 | |||
XM_022418273.1 | XP_022273981.1 | 908 | 140 | |||
XM_022418274.1 | XP_022273982.1 | 1010 | 140 | |||
XM_022418275.1 | XP_022273983.1 | 1009 | 140 | |||
XM_022418276.1 | XP_022273984.1 | 1014 | 140 | |||
XM_022418277.1 | XP_022273985.1 | 525 | 140 | |||
XM_022418280.1 | XP_022273988.1 | 609 | 140 | |||
XM_022418281.1 | XP_022273989.1 | 1013 | 140 | |||
XM_536568.6 | XP_536568.3 | 610 | 140 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
