Assay Details
Gene Symbol: ARHGAP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003434017.4 | XP_003434065.1 | 8 - 9 | 1003 | 146 | |
XM_005639143.3 | XP_005639200.1 | 8 - 9 | 1101 | 146 | ||
XM_005639144.3 | XP_005639201.1 | 6 - 7 | 893 | 146 | ||
XM_022413587.1 | XP_022269295.1 | 6 - 7 | 911 | 146 | ||
XM_022413588.1 | XP_022269296.1 | 5 - 6 | 637 | 146 | ||
XM_535640.6 | XP_535640.4 | 5 - 6 | 687 | 146 | ||
XM_854634.5 | XP_859727.2 | 6 - 7 | 848 | 146 | ||
XM_854701.5 | XP_859794.1 | 6 - 7 | 940 | 146 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
