Assay Details
Gene Symbol: DNAJB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022413467.1 | XP_022269175.1 | 6 - 7 | 808 | 77 | |
XM_544997.6 | XP_544997.1 | 5 - 6 | 830 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
