Assay Details
Gene Symbol: GALNTL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014107585.2 | XP_013963060.1 | 1418 | 130 | ||
XM_022409821.1 | XP_022265529.1 | 1645 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
