Assay Details
| Gene Symbol: KCNIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639596.4 | XP_003639644.2 | 6 - 7 | 696 | 167 | |
| XM_003639597.4 | XP_003639645.1 | 5 - 6 | 532 | 167 | ||
More Information
Set Membership: |
|

KCNIP3
Genomic Map