Assay Details
Gene Symbol: RCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205212.1 | NP_001192141.1 | 4 - 5 | 525 | 113 | |
XM_005617242.3 | XP_005617299.1 | 6 - 7 | 784 | 113 | ||
XM_005617243.3 | XP_005617300.1 | 5 - 6 | 735 | 113 | ||
XM_005617244.3 | XP_005617301.1 | 3 - 4 | 434 | 113 | ||
XM_014106655.2 | XP_013962130.1 | 7 - 8 | 737 | 113 | ||
XM_014106658.2 | XP_013962133.1 | 7 - 8 | 823 | 113 | ||
XM_014106659.2 | XP_013962134.1 | 6 - 7 | 924 | 113 | ||
XM_014106664.2 | XP_013962139.1 | 5 - 6 | 614 | 113 | ||
XM_014106665.2 | XP_013962140.1 | 5 - 6 | 623 | 113 | ||
XM_022405646.1 | XP_022261354.1 | 4 - 5 | 570 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
