Assay Details
| Gene Symbol: CSNK1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625886.3 | XP_005625943.1 | 4 - 5 | 510 | 112 | |
| XM_014117364.2 | XP_013972839.1 | 4 - 5 | 372 | 112 | ||
| XM_022424446.1 | XP_022280154.1 | 4 - 5 | 368 | 112 | ||
| XM_022424447.1 | XP_022280155.1 | 3 - 4 | 337 | 112 | ||
| XM_845075.5 | XP_850168.1 | 3 - 4 | 337 | 112 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CSNK1E
Genomic Map