Assay Details
| Gene Symbol: EIF5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619970.3 | XP_005620027.1 | 3 - 4 | 433 | 98 | |
| XM_005619971.3 | XP_005620028.1 | 3 - 4 | 438 | 98 | ||
| XM_005619972.3 | XP_005620029.1 | 3 - 4 | 415 | 98 | ||
| XM_843935.5 | XP_849028.1 | 3 - 4 | 448 | 98 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

EIF5A
Genomic Map