Assay Details
| Gene Symbol: FCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022423640.1 | XP_022279348.1 | 6 - 7 | 539 | 124 | |
| XM_022423641.1 | XP_022279349.1 | 6 - 7 | 549 | 124 | ||
| XM_845214.3 | XP_850307.1 | 6 - 7 | 517 | 124 | ||
More Information
Set Membership: |
|

FCN2
Genomic Map