Assay Details
| Gene Symbol: GARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639490.4 | XP_003639538.1 | 14 - 15 | 1840 | 141 | |
| XM_014119146.2 | XP_013974621.1 | 14 - 15 | 1662 | 141 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GARS1
Genomic Map