Assay Details
| Gene Symbol: PPP1CA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003064.2 | NP_001003064.2 | 2 - 3 | 284 | 61 | |
| GenBank mRNA | AY062037.1 | - | 2 - 3 | 190 | 61 | |
| DN365927.1 | - | 2 - 3 | 202 | 61 | ||
| DN392813.1 | - | 2 - 3 | 293 | 61 | ||
| DN871864.1 | - | 2 - 3 | 201 | 61 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PPP1CA
Genomic Map