Assay Details
| Gene Symbol: TP63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639826.3 | XP_005639883.1 | 4 - 5 | 801 | 116 | |
| XM_005639827.2 | XP_005639884.1 | 4 - 5 | 688 | 116 | ||
| XM_005639828.3 | XP_005639885.1 | 4 - 5 | 610 | 116 | ||
| XM_022414176.1 | XP_022269884.1 | 4 - 5 | 1103 | 116 | ||
| XM_022414177.1 | XP_022269885.1 | 2 - 3 | 312 | 116 | ||
| XM_856247.5 | XP_861340.3 | 4 - 5 | 1103 | 116 | ||
| XM_856275.3 | XP_861368.1 | 2 - 3 | 427 | 116 | ||
More Information
Set Membership: |
|

TP63
Genomic Map