Assay Details
| Gene Symbol: SEPHS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114736.1 | NP_001108208.1 | 472 | 272 | ||
| GenBank mRNA | DN368851.1 | - | 452 | 272 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SEPHS2
Genomic Map