Assay Details
| Gene Symbol: C29H8orf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195666.1 | NP_001182595.1 | 1 - 2 | 140 | 122 | |
| XM_014109174.2 | XP_013964649.1 | 1 - 2 | 169 | 122 | ||
| XM_022412205.1 | XP_022267913.1 | 1 - 2 | 168 | 122 | ||
| XM_022412206.1 | XP_022267914.1 | 1 - 2 | 168 | 122 | ||
More Information
Set Membership: |
|

C29H8orf34
Genomic Map