Assay Details
Gene Symbol: PRDM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005631306.3 | XP_005631363.1 | 4 - 5 | 698 | 118 | |
XM_005631307.3 | XP_005631364.1 | 4 - 5 | 549 | 118 | ||
XM_005631308.3 | XP_005631365.1 | 4 - 5 | 696 | 118 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
