Assay Details
| Gene Symbol: ZBTB38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014106548.1 | XP_013962023.1 | 2 | 3515 | 149 | |
| XM_022408701.1 | XP_022264409.1 | 3 | 3641 | 149 | ||
| XM_022408702.1 | XP_022264410.1 | 2 | 3480 | 149 | ||
| XM_022408703.1 | XP_022264411.1 | 2 | 3450 | 149 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ZBTB38
Genomic Map