Assay Details
| Gene Symbol: PRPF38B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022420382.1 | XP_022276090.1 | 4 - 5 | 631 | 87 | |
| XM_849600.5 | XP_854693.1 | 2 - 3 | 427 | 87 | ||
More Information
Set Membership: |
|

PRPF38B
Genomic Map