Assay Details
| Gene Symbol: CAMK1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022420861.1 | XP_022276569.1 | 944 | 134 | ||
| XM_547392.6 | XP_547392.3 | 1134 | 134 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CAMK1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022420861.1 | XP_022276569.1 | 944 | 134 | ||
| XM_547392.6 | XP_547392.3 | 1134 | 134 | |||
Set Membership: |
|