Assay Details
Gene Symbol: IMMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630471.3 | XP_005630528.1 | 1 - 2 | 178 | 64 | |
XM_005630472.3 | XP_005630529.1 | 1 - 2 | 178 | 64 | ||
XM_005630473.3 | XP_005630530.1 | 1 - 2 | 178 | 64 | ||
XM_014120431.2 | XP_013975906.1 | 1 - 2 | 178 | 64 | ||
XM_022404742.1 | XP_022260450.1 | 1 - 2 | 178 | 64 | ||
XM_847103.5 | XP_852196.1 | 1 - 2 | 178 | 64 | ||
XM_860208.5 | XP_865301.2 | 1 - 2 | 178 | 64 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
