Assay Details
Gene Symbol: NIF3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005640519.3 | XP_005640576.1 | 2 | 426 | 77 | |
XM_005640520.3 | XP_005640577.1 | 2 | 263 | 77 | ||
XM_005640521.3 | XP_005640578.1 | 2 | 438 | 77 | ||
XM_014110936.2 | XP_013966411.1 | 2 | 284 | 77 | ||
XM_014110937.2 | XP_013966412.1 | 2 | 438 | 77 | ||
XM_014110938.2 | XP_013966413.1 | 2 | 439 | 77 | ||
XM_014110940.2 | XP_013966415.1 | 2 | 438 | 77 | ||
XM_022414985.1 | XP_022270693.1 | 2 | 438 | 77 | ||
XM_844122.5 | XP_849215.1 | 2 | 437 | 77 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
