Assay Details
| Gene Symbol: PRMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616305.3 | XP_005616362.1 | 7 - 8 | 708 | 100 | |
| XM_005616306.3 | XP_005616363.1 | 7 - 8 | 764 | 100 | ||
| XM_533615.6 | XP_533615.2 | 7 - 8 | 827 | 100 | ||
| XM_846450.5 | XP_851543.1 | 8 - 9 | 860 | 100 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PRMT1
Genomic Map