Assay Details
| Gene Symbol: PDLIM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003432137.4 | XP_003432185.1 | 1 - 2 | 228 | 88 | |
| XM_540022.6 | XP_540022.2 | 1 - 2 | 228 | 88 | ||
| XM_843647.5 | XP_848740.1 | 1 - 2 | 227 | 88 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PDLIM3
Genomic Map