Assay Details
Gene Symbol: NDRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284434.1 | NP_001271363.1 | 15 | 1102 | 118 | |
XM_005627974.3 | XP_005628031.1 | 16 | 1195 | 118 | ||
GenBank mRNA | JX987297.1 | - | 15 | 1102 | 118 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
